Tisk…

Voda, LiChrosolv® for chromatography, for LC-MS, Supelco®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

1.15333.1000 1.15333.9185
1.15333.1000EA 639 CZK
1.15333.1000 1.15333.9185
Voda, LiChrosolv® for chromatography, for LC-MS, Supelco®
Voda

LiChrosolv® hypergrade is accurately tested for LC-MS suitability, and meet all the requirements of modern LC-MS ionisation methods (ESI / APCI - positive and negative mode).


Thanks to their low level of ionic background and low ion suppression, LiChrosolv® hypergrade solvents provide high reproducibility and high ionisation efficiency. The packaging material has been improved to perfectly meet LC-MS quality requirements.

Vzorec: H₂O
Bod varu: 100 °C (1013 hPa)
Bod tání: 0 °C
Hustota: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS číslo: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR