Tisk…

Arsenazo III, Millipore®

Dodavatel: Merck
Nebezpečí

Synonyms: 2,7-Bis(2-arsonophenylazo)chromotropic acid, 2,7-Bis(2-arsonophenylazo)-1,8-dihydroxynaphthalene-3,6-disulphonic acid, 2,2′-(1,8-Dihydroxy-3,6-disulphonaphthylene-2,7-bisazo)bisbenzenearsonic acid, Arsenazo Ⅲ

M-Clarity™ quality level = MQ200

1.10107.0005
1.10107.0005EA 38180 CZK
1.10107.0005
Arsenazo III, Millipore®
Arsenazo III
Vzorec: C₂₂H₁₈As₂N₄O₁₄S₂
MW: 776.38
Storage Temperature: Ambient
MDL Number: MFCD00036695
CAS číslo: 1668-00-4
EINECS: 216-788-6
UN: 3465
ADR: 6.1

Objednat

Výsledky specifikačního testu

Solubility (20 °C) slightly soluble
Molar Mass 776.36 g/mol
Bulk Density 850 kg/m3
pH 2.3 (10 g/l, H2O, 20 °C)
Hazard Class 6.1
Hazard Class Description Toxic Materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR