Tisk…

Zinc sulphate 0.1 mol/l concentrated aqueous solution, Titrisol®, Supelco®

Dodavatel: Merck
Nebezpečí


M-Clarity™ quality level = MQ100

1.09991.0001
1.09991.0001EA 1960 CZK
1.09991.0001
Zinc sulphate 0.1 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Zinc sulphate
Vzorec: O₄SZn
Storage Temperature: Ambient
MDL Number: MFCD00011302
CAS číslo: 7733-02-0
EINECS: 231-793-3
UN: 3082
ADR: 9,

Objednat

Výsledky specifikačního testu

Solubility (20 °C) soluble
Density 1.16 g/cm3 (20 °C)
pH 2.2 (H2O, 20 °C)
Hazard Class 9
Hazard Class Description Miscellaneous Hazardous Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR