Tisk…

Etylenglykol 99,5 GC_ASSAY_METHOD, EMSURE® Reag. USP, Reag. Ph. Eur. analysis_for

Dodavatel: Merck
Varování

Synonyms: Ethane-1,2-diol, 1,2-Dihydroxy ethane, 1,2-Ethanediol, Glycol

MQ300

1.09621.1000 1.09621.2500
1.09621.1000EA 2440 CZK
1.09621.1000 1.09621.2500
Etylenglykol 99,5 GC_ASSAY_METHOD, EMSURE® Reag. USP, Reag. Ph. Eur. analysis_for
Etylenglykol

EMSURE® grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE® provides worldwide best and most extensive product specifications. We declare our EMSURE® range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.

Vzorec: HOCH₂CH₂OH
MW: 62.07
Bod varu: 196…198 °C (1013 hPa)
Bod tání: –13 °C
Hustota: 1.115 g/cm³ (20 °C)
Bod vzplanutí: 111 °C
Storage Temperature: Ambient
MDL Number: MFCD00002885
CAS číslo: 107-21-1
EINECS: 203-473-3
REACH: 01-2119456816-28
Merck Index: 13,03832
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4d1c97392851ba184f445f6ee912f84631f98d9&vid=2002300f174929a030d66d9f1394eb6bee89b7dc&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Purity (GC) ≥ 99.5 %
Identity (IR) conforms
Colour ≤ 10 Hazen
Acidity (as CH₃COOH) ≤ 0.001 %
Density (d 20 °C/20 °C) 1.113 - 1.115
Density (d 25 °C/25 °C) 1.10 - 1.12
Refractive index (n 20/D) 1.431 - 1.433
Melting point -14 - -10 °C
Boiling point 196.5 - 198.5 °C
Chloride (Cl) ≤ 0.00002 %
Fe (Iron) ≤ 0.00005 %
Formaldehyde ≤ 0.005 %
Substances reducing potassium permanganate (as O) ≤ 0.0003 %
Readily carbonizable substances conforms
Sulfated ash ≤ 0.005 %
Water ≤ 0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR