Tisk…
pH indicator strips, MQuant®, Supelco®
Dodavatel: Merck
1.09531.0001 1.09532.0001 1.09541.0001 1.09542.0001 1.09543.0001 1.09545.0001 1.09584.0001 1.09450.0010 1.09502.0001
1.09531.0001EA 644 CZK
1.09531.0001 1.09532.0001 1.09541.0001 1.09542.0001 1.09543.0001 1.09545.0001 1.09584.0001 1.09450.0010 1.09502.0001
pH indicator strips, MQuant®, Supelco®
Papírky testovací Stripy pH indikační
These test strips contain special indicator dyes that are covalently bound to the reagent papers which are then adhered to the plastic strips.
  • Non bleeding - do not contaminate the solution to be measured
  • Quality assurance - tests are checked and calibrated against certified buffer solutions
  • Wide and narrow pH ranges available
Informace o príslušenství: Analyse your pH 0 to 14 test strips (1.09535) with this reference card and the free MQuant® StripScan app. The brilliant colour-fast reference colours allow for robust measurement results even underMore Product Information