Tisk…

jod 0.5 mol/l (1 N) aqueous solution, Titripur® Volumetric, Supelco®

Dodavatel: Merck
Nebezpečí


MQ100

1.09098.1000
1.09098.1000EA 4230 CZK
1.09098.1000
jod 0.5 mol/l (1 N) aqueous solution, Titripur® Volumetric, Supelco®
jod
Vzorec: I₂
MW: 253.81
Bod varu: 184.3
Bod tání: 114
Storage Temperature: Ambient
MDL Number: MFCD00011355
CAS číslo: 7553-56-2
EINECS: 231-442-4
UN: 3264
Merck Index: 13,05036

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR