Tisk…

Kyselina trifluoroctová, Uvasol® for spectroscopy, Supelco®

Dodavatel: Merck
Nebezpečí

Synonyms: TFA

M-Clarity™ quality level = MQ100

1.08262.0025 1.08262.0100 1.08262.1000
1.08262.0025EA 1610 CZK
1.08262.0025 1.08262.0100 1.08262.1000
Kyselina trifluoroctová, Uvasol® for spectroscopy, Supelco®
Kyselina trifluoroctová

Accurate analytic results in UV/Vis and infrared spectroscopy depend on the use of very pure solvents for sample preparation. The Uvasol® solvents range has been specially designed for spectroscopy and other applications requiring solvents of the highest spectral purity. The refinement process allows a greater degree of security in applications and avoids misinterpretation of analytical results caused by traces of UV, IR and fluorescence contamination. Uvasol® solvents offer best UV transmittance. In all specifications the minimum transmittance for five typical wavelengths are identified. Furthermore the transmittance is specified in accordance with Reag. Ph. Eur. and ACS.

Vzorec: C₂HF₃O₂
Bod varu: 71,8 °C (1013 hPa)
Bod tání: –15 °C
Hustota: 1,490 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004169
CAS číslo: 76-05-1
EINECS: 200-929-3
UN: 2699
ADR: 8,I
Merck Index: 13,09754
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9501f3ac85d66e8d19d1cd1c3fe5409b974e9d8a&vid=be6aa627129b49ccda76006e7003b46d79b933cc&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Solubility (20 °C) soluble
Melting Point -15 °C
Molar Mass 114.02 g/mol
Boiling Point 72 °C (1013 hPa)
Vapor Pressure 141 hPa (20 °C)
Density 1.48 g/cm³ (20 °C)
pH 1 (10 g/L, H₂O)
Hazard Class 8
Hazard Class Description Corrosive material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR