Tisk…

m-Cresol purple, Supelco®

Dodavatel: Merck

Synonyms: 2',2''-Dimethylphenolsulphonphthalein, m-Cresolsulphonphthalein, MCP, 3,3-Bis(4-hydroxy-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

M-Clarity™ quality level = MQ200

1.05228.0005
1.05228.0005EA 3460 CZK
1.05228.0005
m-Cresol purple, Supelco®
m-Cresol purple
Vzorec: C₂₁H₁₈O₅S
MDL Number: MFCD00005871
CAS číslo: 2303-01-7
EINECS: 218-960-6

Objednat

Výsledky specifikačního testu

Solubility (20 °C) slightly soluble
Molar Mass 382.43 g/mol
Bulk Density 480 kg/m3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR