Tisk…

n-Hexan, EMPLURA®, Supelco®

Dodavatel: Merck
Nebezpečí

Synonyms: Hexyl hydride, Hexanen, Hexane

MQ200

1.04368.1000 1.04368.2500 1.04368.2511
1.04368.1000EA 710 CZK
1.04368.1000 1.04368.2500 1.04368.2511
n-Hexan, EMPLURA®, Supelco®
n-Hexan

EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA® we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA® solvents provide adequate specifications with the most important parameters.

Vzorec: H₃C(CH₂)₄CH₃
MW: 86.18
Bod varu: 69
Bod tání: –94.3 °C
Hustota: 0.659 g/cm³ (20 °C)
Bod vzplanutí: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS číslo: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR