Tisk…
Cellulose for preparative chromatography
Dodavatel: MERCK PRODUCTION CHEMICALS
1.02331.0500
1.02331.0500EA 4720 CZK
1.02331.0500
Cellulose for preparative chromatography
Chromatografii, Sorbenty HPLC sorbenty
Micro-crystalline cellulose is a hydrophilic polysaccharide packing material preferred for the raw separation of amino acids and related compounds. Cellulose is also used for the gentle purification of bio-molecules. Because of the organic nature of cellulose, it can only be used in pre-swollen state and under gentle or hydrostatic pressure.