Tisk…

Petrolether, 40…60 °C, EMSURE® ISO, ACS for analysis, Supelco®

Dodavatel: Merck
Nebezpečí

Synonyms: Gasoline, Petroleum ether, Petrol, Benzine

M-Clarity™ quality level = MQ300

1.01775.1000
1.01775.1000EA 1190 CZK
1.01775.1000
Petrolether, 40…60 °C, EMSURE® ISO, ACS for analysis, Supelco®
Ropný líh

EMSURE® grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications..

Bod varu: 40…60 °C (1013 hPa)
Hustota: 0,65 g/cm³ (20 °C)
Bod vzplanutí: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS číslo: 64742-49-0
EINECS: 265-151-9
UN: 1268
ADR: 3,II

Objednat

Výsledky specifikačního testu

Boiling Point 36 - 83 °C (1013 hPa)
Vapor Pressure 370 hPa (25 °C)
Explosion Limit 1.0 - 7.4 %(V)
Density 0.645 - 0.665 g/cm3 (15 °C)
pH (H2O) not applicable
Viscosity Kinematic 0.45 mm2/s (20 °C)
Grade ACS, ISO


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR