Tisk…
Test kits, fluoride, Spectroquant®, Supelco®
Dodavatel: Merck
1.14598.0001 1.14598.0002 1.00822.0250 1.00809.0001
1.14598.0001EA 5940 CZK
1.14598.0001 1.14598.0002 1.00822.0250 1.00809.0001
Test kits, fluoride, Spectroquant®, Supelco®
Kity pro environmentální testy
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
Spectroquant® cell test kits.
  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells for most the test parameters
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Spectroquant® reagent test kits.
- Highly stable, ready to use reagent mixtures in a user friendly design
- Free selection of appropriate cell format according to the measuring range
- Long shelf life up to 3 years - all at room temperature
- Automatic selection of the correct method with help of AutoSelector barcode identification
Note: The range quoted in the table below refers to the prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.