Tisk…
Organic reference standard, Bisphenol BP, analytical standard, Supelco®
Dodavatel: Merck
08138-100MG
08138-100MGEA 1830 CZK
08138-100MG
Organic reference standard, Bisphenol BP, analytical standard, Supelco®
Standardy Standardy organické
Bisphenol BP belongs to the class of bisphenols.
Bisphenol BP may be used as a reference standard in the determination of bisphenol BP in serum samples using high performance liquid chromatography coupled with tandem mass spectrometry (HPLC-MS/MS).
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=180e10034b4f8137ff6b0555becd830da8b17044&vid=180e10034b4f8137ff6b0555becd830da8b17044&prid=web&clid=SAPDAM