Tisk…

Brilliant green, certified

Dodavatel: VWR Chemicals
Varování

0502-100GEA 9470 CZK
0502-100G
Brilliant green, certified
Brilliant green

A stain for differentation of E.coli from S.typhosa. Also used in bacteriological media. Colour index: 42040

Vzorec: C₂₇H₃₄N₂O₄S
Storage Temperature: Ambient
MDL Number: MFCD00011880
CAS číslo: 633-03-4
EINECS: 211-190-1

Objednat

Výsledky specifikačního testu

Absorbance @ lambda Max ≥0.550
Dye Content ≥85.0%
Lambda max (.003 mg/ml 50% Ethanol) 628 – 632 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR