Tisk…

EDTA dvojsodná sůl dihydrát ≥99.0% for HPLC, Fluka™

Dodavatel: Honeywell Chemicals
Varování

Synonyms: Ethylenediaminetetraacetic acid, disodium salt, dihydrate, Ethylenediaminetetraacetic acid disodium salt dihydrate, Kestranal® 2S dihydrate, Disodium dihydrogen ethylenediaminetetraacetate dihydrate, Ethylenedinitrilotetra-acetic acid disodium salt dihydrate, Ethylenediaminetetraacetic acid disodium dihydrate, Titriplex III, Ethylenediaminetetra-acetic acid disodium salt dihydrate, Kestranal® 2S 2 H2O, Sequestrene Na₂, Edetate disodium dihydrate, Edetic acid disodium salt dihydrate

03682-10G 03682-50G
03682-10GEA 2180 CZK
03682-10G 03682-50G
EDTA dvojsodná sůl dihydrát ≥99.0% for HPLC, Fluka™
EDTA dvojsodná sůl dihydrát
Vzorec: C₁₀H₁₄N₂Na₂O₈·2H₂O
MW: 372.24
Bod tání: 244…250 °C
Storage Temperature: Ambient
MDL Number: MFCD00150037
CAS číslo: 6381-92-6
EINECS: 230-944-0
Merck Index: 13,03543
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Assay 99.0 - 101.0 %
Suitability test (HPLC) complying
Colour of the solution complying
Clearness of the solution complying
Absorbance at 500 nm Max. 0.02
Absorbance at 290 nm Max. 0.02
Absorbance at 280 nm Max. 0.03
Absorbance at 270 nm Max. 0.05
Absorbance at 260 nm Max. 0.15
Aluminium (Al) Max. 5 ppm
Barium (Ba) Max. 5 ppm
Bismuth (Bi) Max. 5 ppm
Calcium (Ca) Max. 10 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 5 ppm
Potassium (K) Max. 50 ppm
Lithium (Li) Max. 5 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Molybdenum (Mo) Max. 5 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Strontium (Sr) Max. 5 ppm
Zinc (Zn) Max. 5 ppm
Total S (as SO₄) Max. 50 ppm
Chloride (Cl) Max. 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR