Tisk…

Acridine orange (hemizinc salt) Electrophoresis

Dodavatel: VWR Chemicals
Nebezpečí

Synonyms: 3,6-Bis(dimethylamino)acridine hydrochloride zinc chloride double salt, Euchrysine 3RX, Acridine orange hemi(Zinc chloride) salt, Acridine orange, Basic orange 14, 3,6-Bis(dimethylamino)acridinium chloride hemi(zinc chloride salt), Euchrysin GNY, N,N,N′,N′-Tetramethylacridine-3,6-diamine monohydrochloride compd. with zinc dichloride

0360-50GEA 0 CZK
0360-50G
Acridine orange (hemizinc salt) Electrophoresis
Acridine orange (hemizinc salt)

A fluorescent stain for nucleic acids in agarose and polyacrylamide gels. Concentrations of 120 ?M will detect DNA at levels of 25-50 ng per band. Also used for to localize nucleic acids in a variety of cell types

Vzorec: C₁₇H₂₀ClN₃·HCl·½ZnCl₂
MW: 369.96 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00081043
CAS číslo: 10127-02-3
EINECS: 233-353-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR