Tisk…

E/E Untaxed 70% water 70 v/v Ph. Helv., Riedel-de Haën™

Dodavatel: Honeywell Chemicals
Nebezpečí

Synonyms: Ethyl alcohol

02877-2.5L 02877-1L
02877-2.5LEA 0 CZK
02877-2.5L 02877-1L
E/E Untaxed 70% water 70 v/v Ph. Helv., Riedel-de Haën™
Ethanol

Upozornení: Untaxed for license holders only

Vzorec: C₂H₅OH
MW: 46.07
Bod varu: 77 °C
Bod tání: -117
Hustota: 0.886 g/cm3
Bod vzplanutí: 22 °C
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS číslo: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3 II
Merck Index: 12,03806

Objednat

Výsledky specifikačního testu

Appearance of the substance complying
Identity (IR) complying
Assay 69.2 - 71.5 Vol.%
Density (D 20/20) 0.883 - 0.889
Non-volatile matter Max. 25 mg/l
Acidity or alkalinity complying
Acetaldehyde & Acetale (GC) Max. 8 Vol.ppm
Benzene (GC) Max. 2 Vol.ppm
Methanol (GC) Max. 150 Vol.ppm
Impurities (GC) total Max. 250 Vol.ppm
Volatile impurities (GC) complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR