Tisk…

Creatine phosphate disodium salt ≥97%, high purity

Dodavatel: VWR Chemicals

Synonyms: Sodium creatine phosphate dibasic, N-Methyl-N-(N-phosphonocarbamimidoyl)glycine, disodium salt, Sodium creatine phosphate, Phosphocreatine disodium salt

0271-25GEA 0 CZK
0271-25G
Creatine phosphate disodium salt ≥97%, high purity
Creatine phosphate disodium salt
Vzorec: C₄H₈N₃O₅P·2Na
Storage Temperature: Freezer
MDL Number: MFCD00014351
CAS číslo: 922-32-7
EINECS: 213-074-6

Objednat

Výsledky specifikačního testu

ATP ≤0.002%
Inorganic Phosphate ≤0.5%
Purity (Enzymatic) >97%
Water (KF) 20 - 24%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR