Tisk…

Jodid draselný, BAKER ANALYZED® ACS, J.T.Baker®

Dodavatel: Avantor

Synonyms: Dipotassium diiodide, Kali iodide

0227.1000EA 17130 CZK
0227.1000 0227.0250 0227.0500
Jodid draselný, BAKER ANALYZED® ACS, J.T.Baker®
Jodid draselný
Vzorec: Kl
MW: 166
Bod varu: 1330
Bod tání: 686
Hustota: 3.13
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS číslo: 7681-11-0
EINECS: 231-659-4
Merck Index: 13,07727

Objednat

Výsledky specifikačního testu

Meets ACS Specifications
Assay min. 99.0 %
Chloride and Bromide (as Cl) max. 0.01 %
Barium (Ba) max. 0.002 %
Calcium (Ca) max. 0.002 %
Magnesium (Mg) max. 0.001 %
Sodium (Na) max. 0.005 %
Insoluble Matter max. 0.005 %
Loss on Drying at 150°C max. 0.2 %
pH of 5% Solution at 25°C 6.0 - 9.2
Phosphate (PO4) max. 0.001 %
Sulfate (SO4) max. 0.005 %
Trace Impurities (in ppm)
Iron (Fe) max. 3
Heavy Metals (as Pb) max. 5
Iodate (IO3) max. 3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR