Tisk…
VWR® Q Path® Formaldehyde Neutraliser
Dodavatel: VWR Chemicals
00699030.EA 3930 CZK
00699030.
VWR® Q Path® Formaldehyde Neutraliser
Prostředky čistící Detergenty a povrchové čističe
Acidic neutraliser and pre-cleaner based on organic acid.
125 g of neutraliser are used to neutralise one L of formalin 4%. Agitate, wait 24 H and the process is completed.