Tisk…

Barium ≥99.2% (metals basis), pieces ≤ 2.5 cm (1 in) (max. 0.8% Sr)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

00653.06 00653.18 00653.30
00653.06EA 2650 CZK
00653.06 00653.18 00653.30
Barium ≥99.2% (metals basis), pieces ≤ 2.5 cm (1 in) (max. 0.8% Sr)
Barium

Purity: 99.2% CAS No.: 7440-39-3 Physical Form: Packaged in oil Appearance: Yellow-silver Odour: Odourless UN No.: 1400 MDL No.: MFCD00134031

Vzorec: Ba
Bod varu: 1640 °C (1013 hPa)
Bod tání: 725 °C
Hustota: 3,51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134031
CAS číslo: 7440-39-3
EINECS: 231-149-1
UN: 1400
ADR: 4.3,II
Merck Index: 12,00991

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR