Kyselina mravenčí 97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Aminic Acid, Methanoic acid
A13285.0C
A13285.0E
A13285.36
A13285.0CEA
5700
CZK
A13285.0C
A13285.0E
A13285.36
Kyselina mravenčí 97%
Kyselina mravenčí
Purity: 97%
CAS No.: 64-18-6
Molecular Formula: CH2O2
Formula Weight: 46.03
Flash Point: 50°(156°F)
Refractive Index: 1.3710
Melting Point: 7-9°
Boiling Point: 100-101°
Beilstein Reference: 1209246
Merck Reference: 14,4241
UN No.: 1779
MDL No.: MFCD00003297
Vzorec:
HCO₂H Bod varu: 100…101 °C Bod tání: 7…9 °C Hustota: 1,220 Bod vzplanutí: 50 °C (156 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003297 CAS číslo: 64-18-6 EINECS: 200-579-1 UN: 1779 ADR: 8,II Merck Index: 14,4241 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...