Tisk…

Butyl cyclopropanesulphonate 95%

Dodavatel: Apollo Scientific

OR952957-1G OR952957-5G
APOSOR952957-1GEA 1830 CZK
APOSOR952957-1G APOSOR952957-5G
Butyl cyclopropanesulphonate 95%
Butyl cyclopropanesulphonate
Vzorec: C₇H₁₄O₃S
Storage Temperature: Ambient
MDL Number: MFCD09261379
CAS číslo: 83635-12-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR