Tisk…

Xanthydrol, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 9-Hydroxyxanthene, 9-Xanthenol, 9H-Xanthen-9-ol

M-Clarity™ quality level = MQ100

252573-25G 252573-5G
SIAL252573-25GEA 5690 CZK
SIAL252573-25G SIAL252573-5G
Xanthydrol, Sigma-Aldrich®
Xanthydrol
Vzorec: C₁₃H₁₀O₂
Bod varu: 338 °C (1013 hPa)
Bod tání: 123 °C
MDL Number: MFCD00005057
CAS číslo: 90-46-0
EINECS: 201-996-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR