Tisk…

Chrom, powder, Sigma-Aldrich®

Dodavatel: Merck

266264-250G 266264-50G
SIAL266264-250GEA 3210 CZK
SIAL266264-250G SIAL266264-50G
Chrom, powder, Sigma-Aldrich®
Chromium
Vzorec: Cr
MDL Number: MFCD00010944
CAS číslo: 7440-47-3
EINECS: 231-157-5
UN: 3089
ADR: 4.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR