3-(4-n-Propoxyphenyl)propionic acid 96%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
H33646.03
H33646.06
H33646.03EA
1320
CZK
H33646.03
H33646.06
3-(4-n-Propoxyphenyl)propionic acid 96%
3-(4-n-Propoxyphenyl)propionic acid
Purity: 96%
CAS No.: 3243-40-1
Molecular Formula: C12H16O3
Formula Weight: 208.26
Melting Point: 94-99°
MDL No.: MFCD00484121
Vzorec:
C₁₂H₁₆O₃ Bod tání: 94…99 °C Storage Temperature: Ambient |
MDL Number:
MFCD00484121 CAS číslo: 3243-40-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...