Tisk…

Dicyklohexylkarbodiimid 1 mol/l in dichloromethane, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 1,3-Dicyclohexylcarbodiimide, DCC, N,N'-Dicyclohexylcarbodiimide

M-Clarity™ quality level = MQ200

379115-100ML 379115-800ML
SIAL379115-100MLEA 1850 CZK
SIAL379115-100ML SIAL379115-800ML
Dicyklohexylkarbodiimid 1 mol/l in dichloromethane, Sigma-Aldrich®
Dicyklohexylkarbodiimid
Vzorec: C₁₃H₂₂N₂
MDL Number: MFCD00011659
CAS číslo: 538-75-0
EINECS: 208-704-1
Merck Index: 13,03123

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR