Tisk…
Anti-CTRP1 (IN) Rabbit Polyclonal Antibody
Dodavatel: ProSci Inc.
3555
PRSI3555EA 12390 CZK
PRSI3555
Anti-CTRP1 (IN) Rabbit Polyclonal Antibody
Protilátky
CTRP1 Antibody: Adipose tissue of an organism plays a major role in regulating physiologic and pathologic processes such as metabolism and immunity by producing and secreting a variety of bioactive molecules termed adipokines. One highly conserved family of adipokines is adiponectin/ACRP30 and its structural and functional paralogs, the C1q/tumor necrosis factor-alpha-related proteins (CTRPs) 1-7. Unlike adiponectin, which is expressed exclusively by differentiated adipocytes, the CTRPs are expressed in a wide variety of tissues. These proteins are thought to act mainly on liver and muscleMore Product Information