Tisk…
Masterflex® Male Luer Slip to Barb Fitting, Avantor®
Dodavatel: Avantor Fluid Handling
MFLX40321-00EA 243 CZK
MFLX40321-00 MFLX40321-01 MFLX40321-02 MFLX40321-03 MFLX40321-04 MFLX40321-05 MFLX40321-06 MFLX40321-07 MFLX40321-08 MFLX40321-09 MFLX40321-10 MFLX40321-11 MFLX40321-12 MFLX40321-13 MFLX40321-14 MFLX40321-15 MFLX40321-16 MFLX40321-17 MFLX40321-18 MFLX40321-19 MFLX40321-20 MFLX40321-21 MFLX40321-22 MFLX40321-23 MFLX40321-24 MFLX40321-25 MFLX40321-26 MFLX40321-27 MFLX40321-28 MFLX40321-29
Masterflex® Male Luer Slip to Barb Fitting, Avantor®
Fitinky a konektory
Easily transition to a luer fitting.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Determine which connector type is needed for your application by contacting free technical application assistance
These fittings are manufactured in an ISO Class 7 cleanroom at an ISO 9001 facility; utilizing an ISO 13485 quality system. There is full lot traceability and tight dimensional tolerances forMore Product Information