Tisk…

2,3,3',4,4',5-Hexachlorobiphenyl (PCB No. 156)

Dodavatel: EHRENSTORFER

C20015600
EHERC20015600EA 3310 CZK
EHERC20015600
2,3,3',4,4',5-Hexachlorobiphenyl (PCB No. 156)
2,3,3',4,4',5-Hexachlorobiphenyl (PCB No. 156)
Vzorec: C₁₂H₄Cl₆
Storage Temperature: Refrigerator
CAS číslo: 38380-08-4
UN: 3432
ADR: 9,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR