Tisk…
Diaphragm vacuum pumps, V-600
Dodavatel: BUCHI
11V600000 11V600010 11V600020 11V600100 11V600110 11V600120 11V600200 11V600210 11V600220 11V600300 11V600310 11V600320
BUCH11V600000EA 131810 CZK
BUCH11V600000 BUCH11V600010 BUCH11V600020 BUCH11V600100 BUCH11V600110 BUCH11V600120 BUCH11V600200 BUCH11V600210 BUCH11V600220 BUCH11V600300 BUCH11V600310 BUCH11V600320
Diaphragm vacuum pumps, V-600
Čerpadla Pumpy vakuové membránové
These powerful and quietly operating three-stage V-600 PTFE diaphragm vacuum pumps are robust and highly flexible. These pumps have been operated at a flow rate of 3,1 m3/h and an ultimate vacuum of 1,5 mbar.
  • High chemical resistant due to the use of high quality materials
  • Easy visual detection of condensate and particles due to transparent front panel
The speed-controlled operation in conjunction with the optional Interface I-300 / I-300 Pro increases the lifetime of the pumps. In combination with the Interface I-300 / I-300 Pro an optional sensorMore Product Information