Tisk…
Anti-CRYG Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Dodavatel: Bioss
BS-13276R-A488
BOSSBS-13276R-A488EA 24950 CZK
BOSSBS-13276R-A488
Anti-CRYG Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Protilátky
Crystallins are the major proteins of the vertebrate eye lens, where they maintain the transparency and refractive index of the lens. Crystallins are divided into alpha, beta, and gamma families, and the beta and gamma-crystallins also comprise a superfamily. Crystallins usually contain seven distinctive protein regions, including four homologous motifs, a connecting peptide, and N- and C-terminal extensions. gamma-crystallins are structural proteins in the lens, and they exists as monomers which typically lack connecting peptides and terminal extensions. The gamma-crystallins include sevenMore Product Information