Tisk…

Anti-IL10 Rabbit Polyclonal Antibody (PE (Phycoerythrin)/Cy3®)

Dodavatel: Biorbyt
ORB129198-100
BIRBORB129198-100EA 0 CZK
BIRBORB129198-100
Anti-IL10 Rabbit Polyclonal Antibody (PE (Phycoerythrin)/Cy3®)
Protilátky
Type: Primary
Antigen: IL10 (interleukin 10)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)/Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR