Tisk…

n-Hexadekan ≥99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Hexadecane, Cetane

A10322.0F A10322.AE A10322.AP
A10322.0FEA 17880 CZK
A10322.0F A10322.AE A10322.AP
n-Hexadekan ≥99%
n-Hexadekan

Hexadecane is use in the recovery of valuable amines by means of distillation from aqueous alkanolamine solutions, which are extensively used in the gas-processing industries.


Purity: 99% CAS No.: 544-76-3 Molecular Formula: CH3(CH2)14CH3 Formula Weight: 226.45 Flash Point: 135°(275°F) Physical Form: Liquid Density: 0.773 Refractive Index: 1.4345 Appearance: Colourless Melting Point: 18° Boiling Point: 287° Solubility: This product is soluble in acetone, alcohol, ether. It is insoluble in water. Beilstein Reference: 1736592 MDL No.: MFCD00008998


Hexadecane is use in the recovery of valuable amines by means of distillation from aqueous alkanolamine solutions, which are extensively used in the gas-processing industries. *Majid Abedinzadegan Abdi and Axel Meisen. Mutual Solubility of Hexadecane with Diethanolamine and with Bis(hydroxyethyl)piperazine J. Chem. Eng. Data, 1998, 43 (2), pp 138-142

Vzorec: H₃C(CH₂)₁₄CH₃
Bod varu: 287 °C
Bod tání: 18 °C
Hustota: 0,773
Bod vzplanutí: 135 °C (275 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008998
CAS číslo: 544-76-3
EINECS: 208-878-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90d3084bdd908b305ecf9b3c30e03cd98a407d94&vid=e5a15f4f6c36024a34521e08083957252ff78457&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR