Tisk…

Chlorid sodný ≥99.8% ISO 9227, ASTM B117 for corrosion testing

Dodavatel: VWR Chemicals

Synonyms: Saline, NaCl, Sea salt, Halite

85139.290EA 1450 CZK
85139.290 85139.360 85139.460
Chlorid sodný ≥99.8% ISO 9227, ASTM B117 for corrosion testing
Chlorid sodný
Vzorec: NaCl
Bod varu: 1413 °C (1013 hPa)
Bod tání: 801 °C
Hustota: 2,16 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS číslo: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Objednat

Výsledky specifikačního testu

Assay ≥ 99.8 %
Anti-caking agent None added
Acidity or alkalinity ≤ 0.0005 meq/g
pH (25°C; 5 %) 5.0 - 7.0
Specific gravity (50 g/l ; 25°C) 1.029 - 1.036
Heavy metals (as Pb) ≤ 2 ppm
Insolubility in water ≤ 50 ppm
Loss on drying (105°C) ≤ 0.1 %
Sodium iodide (as I) ≤ 0.1 %
Total impurities ≤ 0.3 %
Total N (Nitrogen) ≤ 5 ppm
Br (Bromide) ≤ 50 ppm
F (Fluoride) ≤ 0.1 %
I (Iodide) ≤ 20 ppm
Halides (Br, F, I) (Cl excluded) ≤ 0.1 %
PO4 (Phosphate) ≤ 5 ppm
SO4 (Sulphate) ≤ 40 ppm
As (Arsenic) ≤ 0.4 ppm
Ba (Barium) ≤ 10 ppm
Ca (Calcium) ≤ 5 ppm
Cu (Copper) ≤ 0.3 ppm
Fe (Iron) ≤ 1 ppm
K (Potassium) ≤ 10 ppm
Mg (Magnesium) ≤ 2 ppm
Ni (Nickel) ≤ 1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR